Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1410 in Dia, 2 in OAL
Catalog No. 5000515635
Encompass
This Class ZZ Minus Plug & Pin Gage features a diameter of 0.1410 inches and an overall length of 2 inches. It is constructed from steel and comes with a 10 Micro coating/finish, providing precision for various measurement applications.
  • 0.1410 inch diameter
  • 2 inch overall length
  • 0.000200 inch tolerance
  • Steel construction
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • 10 Micro coating/finish
Catalog No. 50-005-15635 Supplier MSC Supplier No. 89014104
This Class ZZ Minus Plug & Pin Gage features a diameter of 0.1410 inches and an overall length of 2 inches. It is constructed from steel and comes with a 10 Micro coating/finish, providing precision for various measurement applications.
  • 0.1410 inch diameter
  • 2 inch overall length
  • 0.000200 inch tolerance
  • Steel construction
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • 10 Micro coating/finish