Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1450 in Dia, 2 in OAL
Catalog No. 5000510854
Encompass
This Class ZZ Minus plug and pin gage is designed for precise measurement and inspection tasks. Made from durable steel, it features a black oxide coating for enhanced longevity and a 0.1450 inch diameter with an overall length of 2 inches.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Ideal for no-go inspection.
  • Constructed from steel for durability.
  • Black oxide coating provides protection.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured in the United States.
Catalog No. 50-005-10854 Supplier MSC Supplier No. 78651650
This Class ZZ Minus plug and pin gage is designed for precise measurement and inspection tasks. Made from durable steel, it features a black oxide coating for enhanced longevity and a 0.1450 inch diameter with an overall length of 2 inches.
  • Used for setting micrometers, verniers, and other measuring devices.
  • Ideal for no-go inspection.
  • Constructed from steel for durability.
  • Black oxide coating provides protection.
  • Includes a certificate of accuracy with NIST traceability.
  • Manufactured in the United States.