Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1540 in, 2 in
Catalog No. 5000534956
Encompass
This Class ZZ Minus Plug & Pin Gage features a diameter of 0.1540 in and an overall length of 2 in. It is constructed from steel with a black oxide finish.
  • Constructed from steel
  • Features a black oxide coating
  • Tolerance of 0.000200 in
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-34956 Supplier MSC Supplier No. 65214710
This Class ZZ Minus Plug & Pin Gage features a diameter of 0.1540 in and an overall length of 2 in. It is constructed from steel with a black oxide finish.
  • Constructed from steel
  • Features a black oxide coating
  • Tolerance of 0.000200 in
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability