Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1585 in Dia, 2 in OAL
Catalog No. 5000530724
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. It is ideal for setting micrometers, conducting NoGo inspections, and use with verniers and other measuring devices. Constructed from steel with a 10 Micro coating/finish, it ensures durability and accuracy.
  • Includes certificate of accuracy with NIST traceability.
  • Made from steel.
  • Features a 10 micro coating/finish.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Measures 0.1585 inch in diameter.
  • Has an overall length of 2 inches.
  • Provides a tolerance of 0.000200 "
Catalog No. 50-005-30724 Supplier MSC Supplier No. 89015853
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. It is ideal for setting micrometers, conducting NoGo inspections, and use with verniers and other measuring devices. Constructed from steel with a 10 Micro coating/finish, it ensures durability and accuracy.
  • Includes certificate of accuracy with NIST traceability.
  • Made from steel.
  • Features a 10 micro coating/finish.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Measures 0.1585 inch in diameter.
  • Has an overall length of 2 inches.
  • Provides a tolerance of 0.000200 "