Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1600 in Dia, 2 in OAL
Catalog No. 5000511096
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. Made from steel with a 10 micro coating/finish, it features a 0.1600 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices. It also includes a Certificate of Accuracy with NIST Traceability.
  • Tolerance of 0.000200 inch
  • Constructed from steel
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
Catalog No. 50-005-11096 Supplier MSC Supplier No. 89016000
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. Made from steel with a 10 micro coating/finish, it features a 0.1600 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices. It also includes a Certificate of Accuracy with NIST Traceability.
  • Tolerance of 0.000200 inch
  • Constructed from steel
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices