Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1840 in
Catalog No. 5000531091
Encompass
This Class ZZ Minus plug and pin gage is designed for precise measurement and inspection in various applications. It offers a specific diameter and overall length, manufactured from durable steel.
  • Measures 0.1840 in diameter
  • Has an overall length of 2 in
  • Constructed from steel
  • Features a 10 Micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers, NoGo inspection, verniers, and other measuring devices
Catalog No. 50-005-31091 Supplier MSC Supplier No. 89018402
This Class ZZ Minus plug and pin gage is designed for precise measurement and inspection in various applications. It offers a specific diameter and overall length, manufactured from durable steel.
  • Measures 0.1840 in diameter
  • Has an overall length of 2 in
  • Constructed from steel
  • Features a 10 Micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers, NoGo inspection, verniers, and other measuring devices