Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.1965 in Diameter, 2 in Length
Catalog No. 5000536930
Encompass
This product is a Class ZZ Minus Plug & Pin Gage with a 0.1965 in diameter and a 2 in overall length. It is constructed from steel and features a 10 Micro finish.
  • Includes certificate of accuracy with NIST traceability.
  • For setting micrometers.
  • For NoGo inspection.
  • For verniers and other measuring devices.
Catalog No. 50-005-36930 Supplier MSC Supplier No. 89019657
This product is a Class ZZ Minus Plug & Pin Gage with a 0.1965 in diameter and a 2 in overall length. It is constructed from steel and features a 10 Micro finish.
  • Includes certificate of accuracy with NIST traceability.
  • For setting micrometers.
  • For NoGo inspection.
  • For verniers and other measuring devices.