Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.2025 in Dia, 2 in OAL
Catalog No. 5000537182
Encompass
This plug and pin gage is designed for precise measurement applications. It features a Class ZZ Minus tolerance, a diameter of 0.2025 in, and an overall length of 2 in.
  • Includes certificate of accuracy with NIST traceability
  • Tolerance of 0.000200 in
  • Features a 10 micro coating/finish
  • Constructed from durable steel
  • Suitable for setting micrometers, NoGo inspection, and other measuring devices
Catalog No. 50-005-37182 Supplier MSC Supplier No. 89020259
This plug and pin gage is designed for precise measurement applications. It features a Class ZZ Minus tolerance, a diameter of 0.2025 in, and an overall length of 2 in.
  • Includes certificate of accuracy with NIST traceability
  • Tolerance of 0.000200 in
  • Features a 10 micro coating/finish
  • Constructed from durable steel
  • Suitable for setting micrometers, NoGo inspection, and other measuring devices