Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.2170 in
Catalog No. 5000515611
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. It features a 0.2170 inch diameter and a 2 inch overall length, making it suitable for no-go inspection and setting various measuring devices. Made from durable steel with a 10 microfinish, it ensures accuracy and includes a certificate of accuracy with NIST traceability.
  • Designed for no-go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Constructed from steel for durability
  • Features a 10 microfinish for precision
  • Includes certificate of accuracy with NIST traceability
  • 0.2170 inch diameter
  • 2 inch overall length
Catalog No. 50-005-15611 Supplier MSC Supplier No. 89021703
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. It features a 0.2170 inch diameter and a 2 inch overall length, making it suitable for no-go inspection and setting various measuring devices. Made from durable steel with a 10 microfinish, it ensures accuracy and includes a certificate of accuracy with NIST traceability.
  • Designed for no-go inspection
  • Suitable for setting micrometers, verniers, and other measuring devices
  • Constructed from steel for durability
  • Features a 10 microfinish for precision
  • Includes certificate of accuracy with NIST traceability
  • 0.2170 inch diameter
  • 2 inch overall length