Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.2200 inch Diameter, 2 inch Overall Length
Catalog No. 5000530474
Encompass
This Class ZZ Minus plug and pin gage features a 0.2200 inch diameter and a 2 inch overall length. It is designed for precise measurement applications, including setting micrometers, performing no-go inspections, and use with verniers and other measuring devices.
  • Features a 0.2200 inch diameter for precise measurements.
  • Has an overall length of 2 inches.
  • Manufactured with a tight tolerance of 0.000200 "
  • Constructed from durable steel for longevity.
  • Finished with black oxide for enhanced protection.
  • Suitable for setting micrometers, no-go inspections, and use with other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed as a Class ZZ Minus gage.
Catalog No. 50-005-30474 Supplier MSC Supplier No. 89022008
This Class ZZ Minus plug and pin gage features a 0.2200 inch diameter and a 2 inch overall length. It is designed for precise measurement applications, including setting micrometers, performing no-go inspections, and use with verniers and other measuring devices.
  • Features a 0.2200 inch diameter for precise measurements.
  • Has an overall length of 2 inches.
  • Manufactured with a tight tolerance of 0.000200 "
  • Constructed from durable steel for longevity.
  • Finished with black oxide for enhanced protection.
  • Suitable for setting micrometers, no-go inspections, and use with other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Designed as a Class ZZ Minus gage.