Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.2830 in Dia, Steel, 10 Micro
Catalog No. 5000515727
Encompass
Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for accurate measurement and inspection tasks. It features a precise diameter, durable steel construction, and a 10 Micro finish for reliability in various applications.
  • Precise diameter of 0.2830 in for accurate measurements.
  • Overall length of 2 in for versatile use.
  • High tolerance of 0.000200 in ensures reliability.
  • Durable steel construction for long-lasting performance.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • 10 Micro finish provides a smooth and accurate surface.
  • Includes a Certificate of Accuracy with NIST Traceability for assured quality.
Catalog No. 50-005-15727 Supplier MSC Supplier No. 89028302
Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for accurate measurement and inspection tasks. It features a precise diameter, durable steel construction, and a 10 Micro finish for reliability in various applications.
  • Precise diameter of 0.2830 in for accurate measurements.
  • Overall length of 2 in for versatile use.
  • High tolerance of 0.000200 in ensures reliability.
  • Durable steel construction for long-lasting performance.
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • 10 Micro finish provides a smooth and accurate surface.
  • Includes a Certificate of Accuracy with NIST Traceability for assured quality.