Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.3065 in, 2 in
Catalog No. 5000515327
Encompass
Vermont Gage Class ZZ Minus Plug & Pin Gages are precision tools designed for accurate measurements in various industrial and laboratory applications. Made from durable tool steel, these gages provide reliable performance for quality control and inspection tasks.
  • Made from durable tool steel with 60/65 Rc hardness.
  • Features Class ZZ tolerance, allowing for .0002 in or .005 mm precision.
  • Standard overall length of 2 in.
  • NIST traceable for guaranteed accuracy.
  • Ideal for measuring hole sizes, slot widths, and performing Go/NoGo gaging.
  • Can be used to size micrometers and snap gages.
Catalog No. 50-005-15327 Supplier MSC Supplier No. 89030654
Vermont Gage Class ZZ Minus Plug & Pin Gages are precision tools designed for accurate measurements in various industrial and laboratory applications. Made from durable tool steel, these gages provide reliable performance for quality control and inspection tasks.
  • Made from durable tool steel with 60/65 Rc hardness.
  • Features Class ZZ tolerance, allowing for .0002 in or .005 mm precision.
  • Standard overall length of 2 in.
  • NIST traceable for guaranteed accuracy.
  • Ideal for measuring hole sizes, slot widths, and performing Go/NoGo gaging.
  • Can be used to size micrometers and snap gages.