Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.3170 Dia, 2 OAL
Catalog No. 5000515991
Encompass
This Class ZZ Minus Plug & Pin Gage is constructed from steel, featuring a precise 0.3170 inch diameter and a 2 inch overall length. Designed with a tolerance of 0.000200 inch and a 10 Micro coating/finish, it is suitable for various precision measurement applications.
  • Includes certificate of accuracy with NIST traceability.
  • Used for setting micrometers.
  • Used for NoGo inspection.
  • Used for verniers and other measuring devices.
Catalog No. 50-005-15991 Supplier MSC Supplier No. 89031702
This Class ZZ Minus Plug & Pin Gage is constructed from steel, featuring a precise 0.3170 inch diameter and a 2 inch overall length. Designed with a tolerance of 0.000200 inch and a 10 Micro coating/finish, it is suitable for various precision measurement applications.
  • Includes certificate of accuracy with NIST traceability.
  • Used for setting micrometers.
  • Used for NoGo inspection.
  • Used for verniers and other measuring devices.