Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.3280 in Dia, 2 in OAL, Steel
Catalog No. 5000537149
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection tasks. It features a 0.3280 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, NoGo inspections, and use with other measuring devices.
  • Constructed from steel
  • Features a 10 Micro finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Tolerance of 0.000200 in
Catalog No. 50-005-37149 Supplier MSC Supplier No. 89032809
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement and inspection tasks. It features a 0.3280 inch diameter and a 2 inch overall length, making it suitable for setting micrometers, NoGo inspections, and use with other measuring devices.
  • Constructed from steel
  • Features a 10 Micro finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Tolerance of 0.000200 in