Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.3375 in, Steel
Catalog No. 5000531862
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage has a 0.3375 inch diameter and an overall length of 2 inches. Constructed from steel, it is suitable for setting micrometers, NoGo inspections, and calibrating verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Features a tolerance of 0.000200 "
  • Coating/finish of 10 micro.
Catalog No. 50-005-31862 Supplier MSC Supplier No. 89033757
This Vermont Gage Class ZZ Minus Plug & Pin Gage has a 0.3375 inch diameter and an overall length of 2 inches. Constructed from steel, it is suitable for setting micrometers, NoGo inspections, and calibrating verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • Features a tolerance of 0.000200 "
  • Coating/finish of 10 micro.