Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.4520 in Diameter, 2 in Overall Length
Catalog No. 5000536896
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Made from steel, it features a 0.4520 in diameter and a 2 in overall length, with a 10 micro coating/finish. It is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Diameter of measurement: 0.4520 in
  • Overall length: 2 in
  • Tolerance: 0.000200 in
  • Steel construction
  • Ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
  • Manufactured in the United States
Catalog No. 50-005-36896 Supplier MSC Supplier No. 89045207
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. Made from steel, it features a 0.4520 in diameter and a 2 in overall length, with a 10 micro coating/finish. It is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Diameter of measurement: 0.4520 in
  • Overall length: 2 in
  • Tolerance: 0.000200 in
  • Steel construction
  • Ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
  • Manufactured in the United States