Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.4525 in Dia, 2 in OAL
Catalog No. 5000511910
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage features a 0.4525 inch diameter and a 2 inch overall length, crafted from durable steel. Designed for precision, it is ideal for various inspection and calibration applications in laboratory and industrial settings.
  • Diameter of 0.4525 inch
  • Overall length of 2 inch
  • Tolerance of 0.000200 inch
  • Constructed from steel
  • Coating/finish of 10 Micro
  • Applicable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-11910 Supplier MSC Supplier No. 89045256
This Vermont Gage Class ZZ Minus Plug & Pin Gage features a 0.4525 inch diameter and a 2 inch overall length, crafted from durable steel. Designed for precision, it is ideal for various inspection and calibration applications in laboratory and industrial settings.
  • Diameter of 0.4525 inch
  • Overall length of 2 inch
  • Tolerance of 0.000200 inch
  • Constructed from steel
  • Coating/finish of 10 Micro
  • Applicable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability