Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.4605 in Dia, 2 in OAL
Catalog No. 5000531464
Encompass
This Class ZZ Minus plug and pin gage, made from steel, features a 0.4605 in diameter and a 2 in overall length. It is designed for precise measurement applications such as setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Features a 0.4605 in diameter
  • Has a 2 in overall length
  • Manufactured from steel
  • Maintains a tolerance of 0.000200 in
  • Finished with 10 microfinish
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-31464 Supplier MSC Supplier No. 89046056
This Class ZZ Minus plug and pin gage, made from steel, features a 0.4605 in diameter and a 2 in overall length. It is designed for precise measurement applications such as setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Features a 0.4605 in diameter
  • Has a 2 in overall length
  • Manufactured from steel
  • Maintains a tolerance of 0.000200 in
  • Finished with 10 microfinish
  • Includes a certificate of accuracy with NIST traceability