Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.4985 in Diameter, 2 in Overall Length, 10 Microfinish
Catalog No. 5000515691
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise NoGo inspection. It features a 0.4985 in diameter and an overall length of 2 in, making it suitable for various measurement applications.
  • Constructed from steel
  • Designed for NoGo inspection applications
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 microfinish coating
  • Has an overall length of 2 in
  • Offers a tolerance of 0.000200 in
  • Measures with a diameter of 0.4985 in
Catalog No. 50-005-15691 Supplier MSC Supplier No. 89049852
This Class ZZ Minus Plug & Pin Gage is designed for precise NoGo inspection. It features a 0.4985 in diameter and an overall length of 2 in, making it suitable for various measurement applications.
  • Constructed from steel
  • Designed for NoGo inspection applications
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 microfinish coating
  • Has an overall length of 2 in
  • Offers a tolerance of 0.000200 in
  • Measures with a diameter of 0.4985 in