Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.5000 in
Catalog No. 5000536455
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool for accurate inspection and calibration. With a 0.5000 inch diameter and 2 inch overall length, it is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Constructed from durable steel with a 10 micro inch coating/finish, it offers enhanced performance and longevity. A Certificate of Accuracy with NIST Traceability is included, ensuring reliable measurements.
  • High accuracy with Class ZZ Minus tolerance of 0.000200 inches.
  • Durable steel construction for long-lasting performance.
  • Optimal 10 micro inch coating/finish for smooth and consistent results.
  • NIST traceability certificate included for reliable and compliant measurements.
  • Versatile for setting micrometers, no-go inspection, and other measuring devices.
Catalog No. 50-005-36455 Supplier MSC Supplier No. 89050009
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool for accurate inspection and calibration. With a 0.5000 inch diameter and 2 inch overall length, it is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Constructed from durable steel with a 10 micro inch coating/finish, it offers enhanced performance and longevity. A Certificate of Accuracy with NIST Traceability is included, ensuring reliable measurements.
  • High accuracy with Class ZZ Minus tolerance of 0.000200 inches.
  • Durable steel construction for long-lasting performance.
  • Optimal 10 micro inch coating/finish for smooth and consistent results.
  • NIST traceability certificate included for reliable and compliant measurements.
  • Versatile for setting micrometers, no-go inspection, and other measuring devices.