Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.5240 in, Steel
Catalog No. 5000510363
Encompass
This steel Class ZZ Minus Plug & Pin Gage, featuring a 0.5240 inch diameter and 2 inch overall length, is designed for precise measurement applications in various settings.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 10 micro coating/finish.
  • Suitable for setting micrometers.
  • Ideal for NoGo inspection.
  • Can be used with verniers and other measuring devices.
  • Constructed from durable steel.
  • Offers a tolerance of 0.000200 in.
Catalog No. 50-005-10363 Supplier MSC Supplier No. 89052401
This steel Class ZZ Minus Plug & Pin Gage, featuring a 0.5240 inch diameter and 2 inch overall length, is designed for precise measurement applications in various settings.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 10 micro coating/finish.
  • Suitable for setting micrometers.
  • Ideal for NoGo inspection.
  • Can be used with verniers and other measuring devices.
  • Constructed from durable steel.
  • Offers a tolerance of 0.000200 in.