Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.5330 in Diameter, 2 in Overall Length
Catalog No. 5000531085
Encompass
This precision steel plug and pin gage is designed for accurate measurement and inspection tasks in various applications. It can be used to set micrometers, perform NoGo inspections, and calibrate verniers and other measuring devices.
  • Diameter of 0.5330 in
  • Overall length of 2 in
  • Tolerance of 0.000200
  • Made from durable steel material
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-31085 Supplier MSC Supplier No. 89053300
This precision steel plug and pin gage is designed for accurate measurement and inspection tasks in various applications. It can be used to set micrometers, perform NoGo inspections, and calibrate verniers and other measuring devices.
  • Diameter of 0.5330 in
  • Overall length of 2 in
  • Tolerance of 0.000200
  • Made from durable steel material
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability