Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.6200 in Dia, 2 in OAL
Catalog No. 5000516014
Encompass
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision instrument designed for accurate measurement and inspection. Constructed from steel, it features a 0.6200 inch diameter and a 2 inch overall length. This gage is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices, and includes a Certificate of Accuracy with NIST Traceability.
  • Constructed from steel
  • Designed for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
  • Provides a tolerance of 0.000200 inch
Catalog No. 50-005-16014 Supplier MSC Supplier No. 89062004
The Vermont Gage Class ZZ Minus Plug & Pin Gage is a precision instrument designed for accurate measurement and inspection. Constructed from steel, it features a 0.6200 inch diameter and a 2 inch overall length. This gage is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices, and includes a Certificate of Accuracy with NIST Traceability.
  • Constructed from steel
  • Designed for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro coating/finish
  • Provides a tolerance of 0.000200 inch