Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.6850 inch Diameter
Catalog No. 5000516186
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise 'NoGo' inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.6850 inch diameter, 2 inch overall length, and is constructed from durable steel with a 10 micro finish. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements.
  • Includes certificate of accuracy with NIST traceability.
  • Ideal for NoGo inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Constructed from steel for durability.
  • Features a 10 micro finish.
Catalog No. 50-005-16186 Supplier MSC Supplier No. 89068506
This Class ZZ Minus Plug & Pin Gage is designed for precise 'NoGo' inspection and for setting micrometers, verniers, and other measuring devices. It features a 0.6850 inch diameter, 2 inch overall length, and is constructed from durable steel with a 10 micro finish. Each gage includes a Certificate of Accuracy with NIST Traceability, ensuring reliable measurements.
  • Includes certificate of accuracy with NIST traceability.
  • Ideal for NoGo inspection.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Constructed from steel for durability.
  • Features a 10 micro finish.