Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.69 mm, 2 in
Catalog No. 5000528886
Encompass
This is a precision plug and pin gage designed for accurate measurement. It features a Class ZZ Minus tolerance, ideal for various inspection and calibration tasks. It measures 0.69 mm in diameter and has a 2 in overall length.
  • Class ZZ Minus tolerance for precise measurements.
  • Measures 0.69 mm in diameter.
  • Features a 2 in overall length.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Accuracy of 10 micro.
Catalog No. 50-005-28886 Supplier MSC Supplier No. 40428146
This is a precision plug and pin gage designed for accurate measurement. It features a Class ZZ Minus tolerance, ideal for various inspection and calibration tasks. It measures 0.69 mm in diameter and has a 2 in overall length.
  • Class ZZ Minus tolerance for precise measurements.
  • Measures 0.69 mm in diameter.
  • Features a 2 in overall length.
  • Suitable for setting micrometers, verniers, and other measuring devices.
  • Accuracy of 10 micro.