Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.7190 in, 2 in
Catalog No. 5000536801
Encompass
The Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection and calibration. Constructed from durable steel with a 10 micro finish, it ensures reliability and longevity. This gage is ideal for setting micrometers, performing NoGo inspections, and verifying other measuring devices, and it comes with a Certificate of Accuracy with NIST Traceability.
  • Precision tool for accurate inspection and calibration.
  • Constructed from durable steel.
  • Features a 10 micro finish for enhanced reliability.
  • Ideal for setting micrometers and NoGo inspections.
  • Suitable for use with verniers and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
Catalog No. 50-005-36801 Supplier MSC Supplier No. 89071906
The Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection and calibration. Constructed from durable steel with a 10 micro finish, it ensures reliability and longevity. This gage is ideal for setting micrometers, performing NoGo inspections, and verifying other measuring devices, and it comes with a Certificate of Accuracy with NIST Traceability.
  • Precision tool for accurate inspection and calibration.
  • Constructed from durable steel.
  • Features a 10 micro finish for enhanced reliability.
  • Ideal for setting micrometers and NoGo inspections.
  • Suitable for use with verniers and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.