Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.7650 in, 2 in
Catalog No. 5000536911
Encompass
This precision gage is a Class ZZ Minus Plug & Pin Gage, meticulously crafted for accurate measurements in various industrial applications. It features a 0.7650 inch diameter and a 2 inch overall length, making it ideal for verifying bore sizes and dimensional accuracy. Constructed from durable steel, this gage is designed for longevity and consistent performance in demanding environments.
  • Class ZZ minus tolerance for precise measurements
  • 0.7650 inch diameter for specific sizing requirements
  • 2 inch overall length for versatile handling
  • Manufactured from durable steel for extended life
  • Includes certificate of accuracy with NIST traceability for assured quality
  • Features a 10 micro coating/finish for enhanced performance
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
Catalog No. 50-005-36911 Supplier MSC Supplier No. 89076509
This precision gage is a Class ZZ Minus Plug & Pin Gage, meticulously crafted for accurate measurements in various industrial applications. It features a 0.7650 inch diameter and a 2 inch overall length, making it ideal for verifying bore sizes and dimensional accuracy. Constructed from durable steel, this gage is designed for longevity and consistent performance in demanding environments.
  • Class ZZ minus tolerance for precise measurements
  • 0.7650 inch diameter for specific sizing requirements
  • 2 inch overall length for versatile handling
  • Manufactured from durable steel for extended life
  • Includes certificate of accuracy with NIST traceability for assured quality
  • Features a 10 micro coating/finish for enhanced performance
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices