Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.8050 in Dia, 2 in OAL, Steel
Catalog No. 5000536762
Encompass
This plug and pin gage is designed for precise measurement applications, featuring a Class ZZ Minus tolerance. It is constructed from steel with a 10 micro finish and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Class ZZ Minus tolerance for precise measurements.
  • Diameter of 0.8050 in.
  • Overall length of 2 in.
  • 10 micro finish for durability.
  • Constructed from steel.
  • Tolerance of 0.000200 in.
  • Includes certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
Catalog No. 50-005-36762 Supplier MSC Supplier No. 89080501
This plug and pin gage is designed for precise measurement applications, featuring a Class ZZ Minus tolerance. It is constructed from steel with a 10 micro finish and includes a Certificate of Accuracy with NIST Traceability, ensuring reliable performance for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.
  • Class ZZ Minus tolerance for precise measurements.
  • Diameter of 0.8050 in.
  • Overall length of 2 in.
  • 10 micro finish for durability.
  • Constructed from steel.
  • Tolerance of 0.000200 in.
  • Includes certificate of accuracy with NIST traceability.
  • Ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices.