Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.8080 in
Catalog No. 5000531679
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. With a diameter of 0.8080 in, an overall length of 2 in, and a tolerance of 0.000200 in, it is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Constructed from steel, it features a 10 micro coating/finish and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for precision measurement
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Constructed from durable steel
  • Features a 10 micro coating/finish
  • Includes a Certificate of Accuracy with NIST Traceability
Catalog No. 50-005-31679 Supplier MSC Supplier No. 89080808
This Class ZZ Minus Plug & Pin Gage is designed for precision measurement applications. With a diameter of 0.8080 in, an overall length of 2 in, and a tolerance of 0.000200 in, it is ideal for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. Constructed from steel, it features a 10 micro coating/finish and includes a Certificate of Accuracy with NIST Traceability.
  • Designed for precision measurement
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Constructed from durable steel
  • Features a 10 micro coating/finish
  • Includes a Certificate of Accuracy with NIST Traceability