Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.8330 in, Steel
Catalog No. 5000515559
Encompass
This steel plug and pin gage features a 0.8330 inch diameter and 2 inch overall length. It is designed for precise applications such as setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • The manufacturer offers a hassle-free guarantee, allowing exchange or refund within 30 days of shipment in original packaging.
  • First quality product.
Catalog No. 50-005-15559 Supplier MSC Supplier No. 89083307
This steel plug and pin gage features a 0.8330 inch diameter and 2 inch overall length. It is designed for precise applications such as setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability.
  • The manufacturer offers a hassle-free guarantee, allowing exchange or refund within 30 days of shipment in original packaging.
  • First quality product.