Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.9020 in, 2 in
Catalog No. 5000536620
Encompass
This Class ZZ Minus Plug & Pin Gage features a 0.9020 in diameter and a 2 in overall length. Constructed from steel with a 10 Micro finish, it is designed for precise measurement applications such as setting micrometers, NoGo inspection, and use with verniers. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ Minus plug and pin gage
  • 0.9020 in diameter
  • 2 in overall length
  • Steel construction
  • 10 Micro coating/finish
  • 0.000200 in tolerance
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
Catalog No. 50-005-36620 Supplier MSC Supplier No. 89090203
This Class ZZ Minus Plug & Pin Gage features a 0.9020 in diameter and a 2 in overall length. Constructed from steel with a 10 Micro finish, it is designed for precise measurement applications such as setting micrometers, NoGo inspection, and use with verniers. Each gage includes a Certificate of Accuracy with NIST Traceability.
  • Class ZZ Minus plug and pin gage
  • 0.9020 in diameter
  • 2 in overall length
  • Steel construction
  • 10 Micro coating/finish
  • 0.000200 in tolerance
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices