Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 0.9650 in Dia, 2 in OAL
Catalog No. 5000515693
Encompass
This steel plug and pin gage is designed for precise measurement applications, featuring a 0.9650 in diameter and a tight tolerance of 0.000200 in. It is suitable for setting micrometers and for "NoGo" inspection with verniers and other measuring devices, offering reliable and verifiable measurements.
  • Ensures high accuracy for critical measurements with a precise diameter and tight tolerance.
  • Made from steel for robustness and longevity.
  • Versatile for setting micrometers and "NoGo" inspection with various measuring devices.
  • Guarantees reliable and verifiable measurements with a certificate of accuracy.
  • Features a 10 micro coating/finish for smooth operation and prolonged tool life.
  • Manufactured in the United States, adhering to quality standards.
Catalog No. 50-005-15693 Supplier MSC Supplier No. 89096507
This steel plug and pin gage is designed for precise measurement applications, featuring a 0.9650 in diameter and a tight tolerance of 0.000200 in. It is suitable for setting micrometers and for "NoGo" inspection with verniers and other measuring devices, offering reliable and verifiable measurements.
  • Ensures high accuracy for critical measurements with a precise diameter and tight tolerance.
  • Made from steel for robustness and longevity.
  • Versatile for setting micrometers and "NoGo" inspection with various measuring devices.
  • Guarantees reliable and verifiable measurements with a certificate of accuracy.
  • Features a 10 micro coating/finish for smooth operation and prolonged tool life.
  • Manufactured in the United States, adhering to quality standards.