Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 1.1505 in Diameter
Catalog No. 5000532115
Encompass
This plug and pin gage is designed for precise measurements, featuring a Class ZZ Minus designation and a diameter of 1.1505 inches. It is suitable for verifying hole sizes and for go/no-go gauging applications.
  • Designed for precise measurements
  • Features a Class ZZ Minus designation
  • Has a diameter of 1.1505 inches
  • Handle sold separately
Catalog No. 50-005-32115 Supplier MSC Supplier No. 64784168
This plug and pin gage is designed for precise measurements, featuring a Class ZZ Minus designation and a diameter of 1.1505 inches. It is suitable for verifying hole sizes and for go/no-go gauging applications.
  • Designed for precise measurements
  • Features a Class ZZ Minus designation
  • Has a diameter of 1.1505 inches
  • Handle sold separately