Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 10.04 mm Dia, 2 inch OAL
Catalog No. 5000529169
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 10.04 mm diameter and a 2 inch overall length, with a tolerance of 0.00508 mm. Constructed from steel, it is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. A Certificate of Accuracy with NIST Traceability is included, and the gage has a 10 Micro coating/finish.
  • Class ZZ Minus plug & pin gage
  • 10.04 mm diameter of measurement
  • 2 inch overall length
  • 0.00508 mm tolerance
  • Constructed from steel
  • Suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • 10 Micro coating/finish
  • Country of origin: United States
Catalog No. 50-005-29169 Supplier MSC Supplier No. 40437493
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications. It features a 10.04 mm diameter and a 2 inch overall length, with a tolerance of 0.00508 mm. Constructed from steel, it is suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices. A Certificate of Accuracy with NIST Traceability is included, and the gage has a 10 Micro coating/finish.
  • Class ZZ Minus plug & pin gage
  • 10.04 mm diameter of measurement
  • 2 inch overall length
  • 0.00508 mm tolerance
  • Constructed from steel
  • Suitable for setting micrometers, NoGo inspection, and use with verniers and other measuring devices
  • Includes a certificate of accuracy with NIST traceability
  • 10 Micro coating/finish
  • Country of origin: United States