Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 12.76 mm Dia, 2 in OAL, Steel
Catalog No. 5000534058
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection. Made from durable steel, it features a 12.76 mm diameter and an overall length of 2 inches, making it suitable for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from durable steel
  • Features a 12.76 mm diameter
  • Measures 2 inches in overall length
  • Suitable for setting micrometers, NoGo inspection, and calibrating measuring devices
  • Coating/finish of 10 micro
  • Tolerance of 0.00508 mm
Catalog No. 50-005-34058 Supplier MSC Supplier No. 40440216
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate inspection. Made from durable steel, it features a 12.76 mm diameter and an overall length of 2 inches, making it suitable for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Includes certificate of accuracy with NIST traceability
  • Manufactured from durable steel
  • Features a 12.76 mm diameter
  • Measures 2 inches in overall length
  • Suitable for setting micrometers, NoGo inspection, and calibrating measuring devices
  • Coating/finish of 10 micro
  • Tolerance of 0.00508 mm