Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 12.93 mm
Catalog No. 5000535438
Encompass
This precision gage is designed for accurate measurement and inspection tasks. It features a Class ZZ Minus tolerance, a 12.93 mm diameter, a 2 inch overall length, and a smooth 10 microinch finish, making it suitable for a wide range of industrial and laboratory applications.
  • Class ZZ minus tolerance
  • 12.93 mm diameter for precise fit
  • 2 inch overall length
  • Smooth 10 microinch finish
  • New condition
Catalog No. 50-005-35438 Supplier MSC Supplier No. 40440380
This precision gage is designed for accurate measurement and inspection tasks. It features a Class ZZ Minus tolerance, a 12.93 mm diameter, a 2 inch overall length, and a smooth 10 microinch finish, making it suitable for a wide range of industrial and laboratory applications.
  • Class ZZ minus tolerance
  • 12.93 mm diameter for precise fit
  • 2 inch overall length
  • Smooth 10 microinch finish
  • New condition