Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 14.63 mm, 2 in Length
Catalog No. 5000511836
Encompass
This Class ZZ Minus Plug & Pin Gage is crafted from steel, featuring a 14.63 mm diameter and a 2 inch overall length. It is designed for precise measurement applications, including 'NoGo' inspection and setting various measuring devices such as micrometers and verniers.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for 'NoGo' inspection.
  • Ideal for setting micrometers and other measuring devices.
  • Constructed from durable steel.
Catalog No. 50-005-11836 Supplier MSC Supplier No. 40442089
This Class ZZ Minus Plug & Pin Gage is crafted from steel, featuring a 14.63 mm diameter and a 2 inch overall length. It is designed for precise measurement applications, including 'NoGo' inspection and setting various measuring devices such as micrometers and verniers.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for 'NoGo' inspection.
  • Ideal for setting micrometers and other measuring devices.
  • Constructed from durable steel.