Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 17.51 mm Dia, Steel
Catalog No. 5000532842
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate verification in various applications. Constructed from durable steel, it features a 17.51 mm diameter and an overall length of 2 inches. It is specifically engineered for tasks such as setting micrometers, conducting no-go inspections, and integrating with verniers and other measuring devices, ensuring high precision in industrial and laboratory settings.
  • Manufactured to Class ZZ Minus specifications for precise measurements.
  • Constructed from durable steel for longevity and reliability.
  • Features a 17.51 mm diameter for specific gaging requirements.
  • Includes a certificate of accuracy with NIST traceability for assured quality.
  • Suitable for setting micrometers, no-go inspection, and other measuring applications.
Catalog No. 50-005-32842 Supplier MSC Supplier No. 40444960
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for accurate verification in various applications. Constructed from durable steel, it features a 17.51 mm diameter and an overall length of 2 inches. It is specifically engineered for tasks such as setting micrometers, conducting no-go inspections, and integrating with verniers and other measuring devices, ensuring high precision in industrial and laboratory settings.
  • Manufactured to Class ZZ Minus specifications for precise measurements.
  • Constructed from durable steel for longevity and reliability.
  • Features a 17.51 mm diameter for specific gaging requirements.
  • Includes a certificate of accuracy with NIST traceability for assured quality.
  • Suitable for setting micrometers, no-go inspection, and other measuring applications.