Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 17.77 mm, 2 in
Catalog No. 5000529553
Encompass
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for setting micrometers, NoGo inspection, verniers, and various other measuring devices. It features a 17.77 mm diameter and a 2 inch overall length, making it suitable for a wide range of laboratory and industrial applications.
  • Constructed from steel for durability
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers
  • Suitable for NoGo inspection
  • Compatible with verniers and other measuring devices
Catalog No. 50-005-29553 Supplier MSC Supplier No. 40445223
This Class ZZ Minus Plug & Pin Gage is a precision measuring tool designed for setting micrometers, NoGo inspection, verniers, and various other measuring devices. It features a 17.77 mm diameter and a 2 inch overall length, making it suitable for a wide range of laboratory and industrial applications.
  • Constructed from steel for durability
  • Features a 10 micro coating/finish
  • Includes a certificate of accuracy with NIST traceability
  • Ideal for setting micrometers
  • Suitable for NoGo inspection
  • Compatible with verniers and other measuring devices