Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 18.21 mm Diameter, 2 in Overall Length, Steel
Catalog No. 5000528141
Encompass
This Class ZZ Minus Plug & Pin Gage features an 18.21 mm diameter of measurement and an overall length of 2 in. It has a tolerance of 0.00508 mm and is constructed from steel with a 10 Micro coating/finish. This gage is designed for applications such as setting micrometers, and for "NoGo" inspection with verniers and other measuring devices. It is manufactured in the United States and includes a Certificate of Accuracy with NIST Traceability.
  • Measures 18.21 mm diameter
  • Overall length of 2 in
  • Features a 0.00508 mm tolerance
  • Constructed from durable steel material
  • Coated with a 10 Micro finish
  • Ideal for setting micrometers
  • Suitable for no-go inspection with verniers and other measuring devices
  • Manufactured in the United States
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-28141 Supplier MSC Supplier No. 40445660
This Class ZZ Minus Plug & Pin Gage features an 18.21 mm diameter of measurement and an overall length of 2 in. It has a tolerance of 0.00508 mm and is constructed from steel with a 10 Micro coating/finish. This gage is designed for applications such as setting micrometers, and for "NoGo" inspection with verniers and other measuring devices. It is manufactured in the United States and includes a Certificate of Accuracy with NIST Traceability.
  • Measures 18.21 mm diameter
  • Overall length of 2 in
  • Features a 0.00508 mm tolerance
  • Constructed from durable steel material
  • Coated with a 10 Micro finish
  • Ideal for setting micrometers
  • Suitable for no-go inspection with verniers and other measuring devices
  • Manufactured in the United States
  • Includes a certificate of accuracy with NIST traceability