Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 19.16 mm Diameter, 50.8 mm Length, Steel
Catalog No. 5000536818
Encompass
The Class ZZ Minus Plug & Pin Gage is a precision measurement tool designed for various inspection applications. It features a 19.16 mm diameter and an overall length of 50.8 mm, constructed from durable steel.
  • Designed for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 10 micro coating/finish.
  • Constructed from steel for durability.
  • Measurement diameter of 19.16 mm.
  • Overall length of 50.8 mm (2 inches).
  • Provides a tolerance of 0.00508 mm.
Catalog No. 50-005-36818 Supplier MSC Supplier No. 40446619
The Class ZZ Minus Plug & Pin Gage is a precision measurement tool designed for various inspection applications. It features a 19.16 mm diameter and an overall length of 50.8 mm, constructed from durable steel.
  • Designed for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a 10 micro coating/finish.
  • Constructed from steel for durability.
  • Measurement diameter of 19.16 mm.
  • Overall length of 50.8 mm (2 inches).
  • Provides a tolerance of 0.00508 mm.