Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 19.25 mm, 2 in
Catalog No. 5000529055
Encompass
This precision gage is designed for accurate measurement applications, including setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. Crafted from durable steel, it features a 19.25 mm diameter and a 2 inch overall length, ensuring reliability and consistent performance. Each gage includes a certificate of accuracy with NIST traceability for verified precision.
  • Class ZZ minus tolerance
  • 19.25 mm diameter
  • 2 inch overall length
  • 0.00508 mm tolerance
  • Steel material
  • 10 micro coating/finish
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-29055 Supplier MSC Supplier No. 40446700
This precision gage is designed for accurate measurement applications, including setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. Crafted from durable steel, it features a 19.25 mm diameter and a 2 inch overall length, ensuring reliability and consistent performance. Each gage includes a certificate of accuracy with NIST traceability for verified precision.
  • Class ZZ minus tolerance
  • 19.25 mm diameter
  • 2 inch overall length
  • 0.00508 mm tolerance
  • Steel material
  • 10 micro coating/finish
  • Includes certificate of accuracy with NIST traceability