Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 19.78 mm Dia, 2 in OAL
Catalog No. 5000527045
Encompass
This Class ZZ Minus Plug & Pin Gage from Vermont Gage is designed for precise measurement and inspection. With a diameter of 19.78 mm and an overall length of 2 inches, it offers a tight tolerance of 0.00508 mm. Constructed from durable steel with a 10 micro finish, this gage is suitable for setting micrometers, performing NoGo inspection, and calibrating verniers and other measuring devices.
  • 19.78 mm diameter measurement
  • 2 inch overall length
  • 0.00508 mm tolerance
  • Steel material
  • 10 micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-27045 Supplier MSC Supplier No. 40447237
This Class ZZ Minus Plug & Pin Gage from Vermont Gage is designed for precise measurement and inspection. With a diameter of 19.78 mm and an overall length of 2 inches, it offers a tight tolerance of 0.00508 mm. Constructed from durable steel with a 10 micro finish, this gage is suitable for setting micrometers, performing NoGo inspection, and calibrating verniers and other measuring devices.
  • 19.78 mm diameter measurement
  • 2 inch overall length
  • 0.00508 mm tolerance
  • Steel material
  • 10 micro coating/finish
  • Suitable for setting micrometers, NoGo inspection, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability