Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 2.88 mm, 2 in OAL, Steel
Catalog No. 5000513968
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications such as setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. Featuring a 2.88 mm diameter and a 2-inch overall length, this steel gage offers reliable performance for various inspection needs. Each gage includes a Certificate of Accuracy with NIST Traceability to ensure precision.
  • Designed for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Constructed from steel for durability.
  • Features a Class ZZ Minus tolerance.
  • Finished with a 10 micro coating.
Catalog No. 50-005-13968 Supplier MSC Supplier No. 40430332
This Vermont Gage Class ZZ Minus Plug & Pin Gage is designed for precise measurement applications such as setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. Featuring a 2.88 mm diameter and a 2-inch overall length, this steel gage offers reliable performance for various inspection needs. Each gage includes a Certificate of Accuracy with NIST Traceability to ensure precision.
  • Designed for setting micrometers, NoGo inspection, verniers, and other measuring devices.
  • Includes a Certificate of Accuracy with NIST Traceability.
  • Constructed from steel for durability.
  • Features a Class ZZ Minus tolerance.
  • Finished with a 10 micro coating.