Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 21.26 mm, 2 in, Steel
Catalog No. 5000512265
Encompass
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. It features a 21.26 mm diameter and a 2 inch overall length. Constructed from steel, it is suitable for various applications requiring high accuracy.
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Made from durable steel material
  • Features a 10 micro coating/finish
Catalog No. 50-005-12265 Supplier MSC Supplier No. 40448714
This Class ZZ Minus Plug & Pin Gage is designed for precise measurement and inspection tasks. It features a 21.26 mm diameter and a 2 inch overall length. Constructed from steel, it is suitable for various applications requiring high accuracy.
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers
  • Ideal for NoGo inspection
  • Compatible with verniers and other measuring devices
  • Made from durable steel material
  • Features a 10 micro coating/finish