Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 21.73 mm, 2 in OAL
Catalog No. 5000512659
Encompass
The Class ZZ Minus Plug & Pin Gage has a 21.73 mm diameter and a 2 in overall length. This precision instrument is designed for accurate measurement and inspection tasks in various applications.
  • Used to set micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection.
  • Manufactured from steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a tolerance of 0.00508 mm.
  • Coating provides a 10 micro finish.
Catalog No. 50-005-12659 Supplier MSC Supplier No. 40449183
The Class ZZ Minus Plug & Pin Gage has a 21.73 mm diameter and a 2 in overall length. This precision instrument is designed for accurate measurement and inspection tasks in various applications.
  • Used to set micrometers, verniers, and other measuring devices.
  • Suitable for NoGo inspection.
  • Manufactured from steel.
  • Includes a certificate of accuracy with NIST traceability.
  • Features a tolerance of 0.00508 mm.
  • Coating provides a 10 micro finish.