Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 3.18 mm Diameter, 2 in Overall Length
Catalog No. 5000534045
Encompass
This Vermont Gage Class ZZ Minus Plug & Pin Gage offers precise measurement with a 3.18 mm diameter and 2 in overall length. Crafted from robust tool steel with Rc 60/65 hardness and a 10 microfinish, it's ideal for setting micrometers, verniers, and for NoGo inspection. Each gage provides a Certificate of Accuracy with NIST Traceability for assured reliability.
  • Class ZZ minus plug and pin gage
  • 3.18 mm diameter
  • 2 in overall length
  • 0.00508 mm tolerance
  • Tool steel with Rc 60/65 hardness
  • 10 microfinish
  • For setting micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
  • Includes Certificate of Accuracy with NIST Traceability
  • Made in the United States
Catalog No. 50-005-34045 Supplier MSC Supplier No. 40430639
This Vermont Gage Class ZZ Minus Plug & Pin Gage offers precise measurement with a 3.18 mm diameter and 2 in overall length. Crafted from robust tool steel with Rc 60/65 hardness and a 10 microfinish, it's ideal for setting micrometers, verniers, and for NoGo inspection. Each gage provides a Certificate of Accuracy with NIST Traceability for assured reliability.
  • Class ZZ minus plug and pin gage
  • 3.18 mm diameter
  • 2 in overall length
  • 0.00508 mm tolerance
  • Tool steel with Rc 60/65 hardness
  • 10 microfinish
  • For setting micrometers, verniers, and other measuring devices
  • Suitable for NoGo inspection
  • Includes Certificate of Accuracy with NIST Traceability
  • Made in the United States