Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 3.89 mm Dia, 2 in OAL
Catalog No. 5000527855
Encompass
This Class ZZ Minus Plug & Pin Gage features a 3.89 mm diameter and a 2 inch overall length, designed for precise measurement and calibration. Constructed from durable steel, it is ideal for NoGo inspection and for accurately setting micrometers, verniers, and other measuring devices.
  • Features a 10 microfinish for enhanced performance.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for precise NoGo inspection.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Manufactured from durable steel.
  • Meets ASME B89.1 standards.
Catalog No. 50-005-27855 Supplier MSC Supplier No. 40431348
This Class ZZ Minus Plug & Pin Gage features a 3.89 mm diameter and a 2 inch overall length, designed for precise measurement and calibration. Constructed from durable steel, it is ideal for NoGo inspection and for accurately setting micrometers, verniers, and other measuring devices.
  • Features a 10 microfinish for enhanced performance.
  • Includes a certificate of accuracy with NIST traceability.
  • Suitable for precise NoGo inspection.
  • Designed for setting micrometers, verniers, and other measuring devices.
  • Manufactured from durable steel.
  • Meets ASME B89.1 standards.