Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 4.42 mm, 2 in
Catalog No. 5000514070
Encompass
The Vermont Gage Class ZZ Minus Plug and Pin Gage is a precision instrument for accurate measurement and inspection. With a 4.42 mm diameter and 2-inch overall length, this steel gage suits various laboratory and industrial applications. It ensures reliable results for setting measurement devices and Go/NoGo inspections.
  • Designed for quick and accurate measurements.
  • Suitable for Go/NoGo inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Gages over 0.0605 inch are marked with diameter and tolerance.
  • Enables gaging of slot widths and locations.
  • Includes a certificate of accuracy.
  • Constructed from tool steel with 60/65 Rc hardness.
  • Features Class ZZ tolerance (0.005 mm).
  • Has a 10 microinch finish or better.
Catalog No. 50-005-14070 Supplier MSC Supplier No. 40431876
The Vermont Gage Class ZZ Minus Plug and Pin Gage is a precision instrument for accurate measurement and inspection. With a 4.42 mm diameter and 2-inch overall length, this steel gage suits various laboratory and industrial applications. It ensures reliable results for setting measurement devices and Go/NoGo inspections.
  • Designed for quick and accurate measurements.
  • Suitable for Go/NoGo inspection.
  • Can be used to set micrometers, verniers, and other measuring devices.
  • Gages over 0.0605 inch are marked with diameter and tolerance.
  • Enables gaging of slot widths and locations.
  • Includes a certificate of accuracy.
  • Constructed from tool steel with 60/65 Rc hardness.
  • Features Class ZZ tolerance (0.005 mm).
  • Has a 10 microinch finish or better.