Learn More
MSC Vermont Gage Plug & Pin Gage, Class ZZ Minus, 4.53 mm, 2 in
Catalog No. 5000532727
Encompass
This Class ZZ Minus plug and pin gage is designed for precise measurement applications, including setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. Constructed from steel, it features a 4.53 mm diameter, a 2 inch overall length, and a tolerance of 0.00508 mm, ensuring accuracy for critical inspections.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from durable steel for longevity.
  • Features a 4.53 mm diameter for precise measurements.
  • Measures 2 inch in overall length.
  • Designed with a tolerance of 0.00508 mm.
  • Ideal for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Equipped with a 10 micro coating/finish.
Catalog No. 50-005-32727 Supplier MSC Supplier No. 40431983
This Class ZZ Minus plug and pin gage is designed for precise measurement applications, including setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices. Constructed from steel, it features a 4.53 mm diameter, a 2 inch overall length, and a tolerance of 0.00508 mm, ensuring accuracy for critical inspections.
  • Includes certificate of accuracy with NIST traceability.
  • Constructed from durable steel for longevity.
  • Features a 4.53 mm diameter for precise measurements.
  • Measures 2 inch in overall length.
  • Designed with a tolerance of 0.00508 mm.
  • Ideal for setting micrometers, NoGo inspection, and calibrating verniers and other measuring devices.
  • Equipped with a 10 micro coating/finish.